Assay Details
Target Gene Details
Entrez Gene ID: | 8501 |
Gene Name: | solute carrier family 43 member 1 |
Gene Aliases: |
LAT3, PB39, POV1, R00504 |
Location: |
Chr.11:57484531-57515954 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC43A1 | NM_001198810.1 | NP_001185739.1 | ||
NM_003627.5 | NP_003618.1 | |||
XM_005274358.4 | XP_005274415.1 | |||
XM_011545320.2 | XP_011543622.1 | |||
XM_011545321.2 | XP_011543623.1 | |||
XM_011545322.1 | XP_011543624.1 | |||
XM_017018451.1 | XP_016873940.1 | |||
XM_017018452.1 | XP_016873941.1 | |||
XM_017018453.1 | XP_016873942.1 | |||
AB103033.1 | BAD00152.1 | |||
AF045584.1 | AAC33004.1 | |||
AK027732.1 | ||||
BC001639.2 | AAH01639.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |