Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_001318380.1 | NP_001305309.1 | ||
NM_177533.4 | NP_803877.2 | |||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | ||||
AK310449.1 | ||||
BC041584.1 | AAH41584.1 | |||
DA249638.1 | ||||
DA409636.1 | ||||
DB096906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv566177 | Chr.14:105167384 - 105177639 on Build GRCh38 | Gain | JAG2 NUDT14 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | MTA1 BTBD6 BRF1 GPR132 JAG2 MIR6765 CRIP2 LOC100507437 TEX22 PACS2 NUDT14 LOC102723354 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | MTA1 IGH BTBD6 C14orf80 NUDT14 TMEM121 CRIP1 BRF1 LOC105370697 JAG2 MIR6765 CRIP2 LOC100507437 TEX22 PACS2 IGHA2 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | BRF1 GPR132 JAG2 MIR6765 NUDT14 LOC102723354 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | BRF1 JAG2 MIR6765 NUDT14 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | BTBD6 KIF26A KLC1 ZFYVE21 C14orf79 BAG5 ASPG NUDT14 LOC105370691 ZBTB42 CKB LINC00638 PLD4 SIVA1 MIR203B INF2 LINC00637 AHNAK2 PACS2 RD3L CDCA4 MIR4710 LOC102723354 C14orf180 TRMT61A APOPT1 MIR203A ADSSL1 LOC388022 LOC101929634 LOC107984648 MARK3 LOC105370708 LOC105370687 CEP170B C14orf2 XRCC3 LOC105378183 SNORA28 BRF1 GPR132 TMEM179 JAG2 EIF5 MIR6765 TDRD9 PPP1R13B AKT1 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | BTBD6 BRF1 GPR132 JAG2 MIR6765 C14orf79 AHNAK2 NUDT14 CDCA4 LOC102723354 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | MIR4539 IGHA1 ELK2AP MIR4538 IGHV3-23 BTBD6 MIR4507 IGHG3 ADAM6 C14orf80 NUDT14 TMEM121 IGHG1 MIR8071-2 IGHD MIR4537 MIR8071-1 TEX22 PACS2 IGHV3-48 IGHV3-41 IGHV4-55 IGHM MTA1 IGH LOC107987211 IGHG4 IGHV1-69 KIAA0125 IGHG2 CRIP1 IGHV3-71 LINC00221 BRF1 LOC105370697 LINC00226 JAG2 MIR6765 IGHV3-69-1 IGHV3-30 CRIP2 LOC100507437 IGHV4-31 IGHA2 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | BRF1 GPR132 JAG2 MIR6765 C14orf79 AHNAK2 NUDT14 CDCA4 LOC102723354 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |