Assay Details
Target Gene Details
Entrez Gene ID: | 83707 |
Gene Name: | tRNA phosphotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.11:64223799-64226254 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRPT1 | NM_001033678.3 | NP_001028850.2 | ||
NM_001160389.1 | 4 | 514 | NP_001153861.1 | |
NM_001160390.1 | NP_001153862.1 | |||
NM_001160392.1 | NP_001153864.1 | |||
NM_001160393.1 | NP_001153865.1 | |||
NM_031472.3 | NP_113660.1 | |||
XM_005274345.4 | XP_005274402.1 | |||
XM_005274346.4 | XP_005274403.1 | |||
XM_005274347.4 | XP_005274404.1 | |||
XM_006718706.3 | XP_006718769.1 | |||
XM_006718707.3 | XP_006718770.1 | |||
XM_017018400.1 | XP_016873889.1 | |||
XM_017018401.1 | XP_016873890.1 | |||
AK026364.1 | ||||
AW771318.1 | ||||
AY211494.1 | AAO62941.1 | |||
BC005133.2 | AAH05133.1 | |||
BG105545.1 | ||||
BM982538.1 | ||||
BQ672032.1 | ||||
BQ674339.1 | ||||
DR004975.1 | 4 | 412 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)