Assay Details
Target Gene Details
Entrez Gene ID: | 5354 |
Gene Name: | proteolipid protein 1 |
Gene Aliases: |
GPM6C, HLD1, MMPL, PLP, PLP/DM20, PMD, SPG2 |
Location: |
Chr.X:103776506-103792619 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLP1 | NM_000533.4 | 7 | 1930 | NP_000524.3 |
NM_001128834.2 | 8 | 2012 | NP_001122306.1 | |
NM_001305004.1 | 7 | 1705 | NP_001291933.1 | |
NM_199478.2 | 7 | 1825 | NP_955772.1 | |
AI205189.1 | 1 | 304 | ||
AK128782.1 | 7 | 1853 | ||
BC002665.1 | 7 | 1768 | AAH02665.1 | |
BC095452.1 | 7 | 1814 | AAH95452.1 | |
BQ723748.1 | 1 | 645 | ||
EB387051.1 | 1 | 170 | ||
M27110.1 | 7 | 1845 | AAA60117.1 | |
M54927.1 | 7 | 1848 | AAA59565.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |