Assay Details
Target Gene Details
Entrez Gene ID: | 7398 |
Gene Name: | ubiquitin specific peptidase 1 |
Gene Aliases: |
UBP |
Location: |
Chr.1:62436304-62451804 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP1 | NM_001017415.1 | NP_001017415.1 | ||
NM_001017416.1 | NP_001017416.1 | |||
NM_003368.4 | 1 | 475 | NP_003359.3 | |
AB208893.1 | 1 | 107 | BAD92130.1 | |
AF117386.1 | AAD11441.1 | |||
AK312423.1 | 1 | 138 | ||
AL599240.1 | ||||
BC018745.1 | 1 | 475 | ||
BC032364.1 | 1 | 125 | ||
BC050525.1 | 1 | 179 | AAH50525.1 | |
BI463072.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |