Assay Details
Target Gene Details
Entrez Gene ID: | 169792 |
Gene Name: | GLIS family zinc finger 3 |
Gene Aliases: |
NDH, ZNF515 |
Location: |
Chr.9:3824127-4310694 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLIS3 | NM_001042413.1 | 11 | 3509 | NP_001035878.1 |
NM_152629.3 | 10 | 2525 | NP_689842.3 | |
XM_005251386.4 | 10 | 3068 | XP_005251443.1 | |
XM_005251387.4 | 10 | 3346 | XP_005251444.1 | |
XM_005251388.4 | 9 | 2860 | XP_005251445.1 | |
XM_011517763.2 | 11 | 3188 | XP_011516065.1 | |
XM_011517764.2 | 11 | 3018 | XP_011516066.1 | |
XM_011517766.2 | 10 | 3597 | XP_011516068.1 | |
XM_011517767.2 | 10 | 2991 | XP_011516069.1 | |
XM_017014361.1 | 10 | 3296 | XP_016869850.1 | |
AB209404.1 | 8 | 1118 | BAD92641.1 | |
AK075059.1 | ||||
AK096318.1 | 8 | 1152 | ||
BC033899.2 | 10 | 2525 | AAH33899.2 | |
DQ438878.1 | 8 | 1453 | ABE66435.1 | |
KU178885.1 | ||||
KU178886.1 | ||||
KU178887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1028973 | Chr.9:3789264 - 3856678 on Build GRCh38 | Loss | GLIS3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |