Assay Details
Target Gene Details
Entrez Gene ID: | 2900 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 4 |
Gene Aliases: |
EAA1, GRIK, GluK4, KA1 |
Location: |
Chr.11:120511746-120988805 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIK4 | NM_001282470.2 | NP_001269399.1 | ||
NM_001282473.2 | NP_001269402.1 | |||
NM_014619.4 | NP_055434.2 | |||
XM_011542784.1 | XP_011541086.1 | |||
XM_011542786.2 | XP_011541088.1 | |||
XM_011542787.2 | XP_011541089.1 | |||
XM_017017621.1 | XP_016873110.1 | |||
XM_017017622.1 | XP_016873111.1 | |||
AK292726.1 | ||||
AK314284.1 | ||||
BC146652.1 | ||||
BC150173.1 | ||||
S67803.1 | AAB29311.1 |
Target Gene Details
Entrez Gene ID: | 101929208 |
Gene Name: | uncharacterized LOC101929208 |
Gene Aliases: |
- |
Location: |
Chr.11:120940554-120958039 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929208 | NR_133004.1 | |||
BX107382.1 | ||||
HY049706.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |