Assay Details
Target Gene Details
Entrez Gene ID: | 63908 |
Gene Name: | NSF attachment protein beta |
Gene Aliases: |
SNAP-BETA, SNAPB |
Location: |
Chr.20:23374519-23421519 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAPB | NM_001283018.1 | 11 | 2748 | NP_001269947.1 |
NM_001283020.1 | 10 | 2619 | NP_001269949.1 | |
NM_001283026.1 | 10 | 2689 | NP_001269955.1 | |
NM_022080.2 | 11 | 2736 | NP_071363.1 | |
NR_104266.1 | ||||
XM_011529313.1 | 11 | 2740 | XP_011527615.1 | |
XM_017028008.1 | 11 | 2943 | XP_016883497.1 | |
AK022817.1 | 7 | 2262 | ||
AK124876.1 | 11 | 2675 | ||
AK125811.1 | 4 | 3595 | ||
AK126340.1 | 7 | 4086 | ||
AL834445.1 | 5 | 2152 | CAD39105.1 | |
BC047426.1 | 10 | 2631 | AAH47426.1 | |
BC060840.1 | 11 | 2661 | AAH60840.1 | |
BX648330.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |