Assay Details
Target Gene Details
Entrez Gene ID: | 9797 |
Gene Name: | TatD DNase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:10248493-10281222 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TATDN2 | NM_014760.3 | 4 | 1998 | NP_055575.3 |
AK289794.1 | 4 | 1719 | ||
BC090935.1 | 4 | 1579 | AAH90935.1 | |
BC101770.1 | 4 | 1487 | AAI01771.1 | |
BC101776.1 | 4 | 1488 | AAI01777.1 | |
D86972.1 | 5 | 1815 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | ATP2B2 SLC6A1-AS1 FANCD2OS BRK1 FANCD2 MIR378B LINC00852 IRAK2 SLC6A11 LOC101927467 LINC00606 VGLL4 GHRL VHL SLC6A1 MIR885 TAMM41 GHRLOS ATP2B2-IT2 ATG7 SEC13 TATDN2 HRH1 |
nsv589650 | Chr.3:10260488 - 10294102 on Build GRCh38 | Loss | GHRL GHRLOS LINC00852 TATDN2 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | ATP2B2 SLC6A1-AS1 FANCD2 EMC3 MIR378B IRAK2 SLC6A11 LINC00606 VGLL4 VHL GHRLOS CIDECP ATP2B2-IT2 SEC13 TATDN2 FANCD2OS BRK1 FANCD2P2 LINC00852 LOC101927467 LOC401052 GHRL SLC6A1 MIR885 TAMM41 ATG7 HRH1 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | ATP2B2 SLC6A1-AS1 FANCD2OS BRK1 FANCD2 FANCD2P2 MIR378B LINC00852 IRAK2 SLC6A11 LOC101927467 LINC00606 VGLL4 GHRL VHL SLC6A1 MIR885 TAMM41 GHRLOS ATP2B2-IT2 ATG7 SEC13 TATDN2 HRH1 |
nsv236904 | Chr.3:10266949 - 10274997 on Build GRCh38 | Deletion | TATDN2 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | ATP2B2 SLC6A1-AS1 FANCD2OS BRK1 FANCD2 FANCD2P2 MIR378B LINC00852 IRAK2 SLC6A11 LOC101927467 LINC00606 VGLL4 GHRL VHL SLC6A1 MIR885 TAMM41 GHRLOS ATP2B2-IT2 ATG7 SEC13 TATDN2 HRH1 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | ATP2B2 SLC6A1-AS1 FANCD2 EMC3 MIR378B IRAK2 SLC6A11 LINC00606 VGLL4 VHL GHRLOS CIDECP ATP2B2-IT2 SEC13 TATDN2 FANCD2OS BRK1 FANCD2P2 LINC00852 LOC101927467 LOC401052 GHRL SLC6A1 MIR885 TAMM41 ATG7 EMC3-AS1 HRH1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |