Assay Details
Target Gene Details
Entrez Gene ID: | 4798 |
Gene Name: | nuclear factor related to kappaB binding protein |
Gene Aliases: |
INO80G |
Location: |
Chr.11:129863773-129895596 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFRKB | NM_001143835.1 | 22 | 2598 | NP_001137307.1 |
NM_006165.3 | 20 | 2593 | NP_006156.2 | |
XM_005271567.3 | 22 | 2634 | XP_005271624.1 | |
XM_005271568.3 | 21 | 2549 | XP_005271625.1 | |
XM_011542850.2 | 23 | 2671 | XP_011541152.1 | |
XM_011542851.2 | 22 | 2653 | XP_011541153.1 | |
XM_011542852.2 | 21 | 2491 | XP_011541154.1 | |
XM_017017793.1 | 21 | 2545 | XP_016873282.1 | |
XM_017017794.1 | 21 | 2509 | XP_016873283.1 | |
XM_017017795.1 | 23 | 2635 | XP_016873284.1 | |
XM_017017796.1 | 21 | 2513 | XP_016873285.1 | |
XM_017017797.1 | 21 | 2545 | XP_016873286.1 | |
XM_017017798.1 | 22 | 2598 | XP_016873287.1 | |
XM_017017799.1 | 23 | 2745 | XP_016873288.1 | |
XM_017017800.1 | 23 | 2685 | XP_016873289.1 | |
XM_017017801.1 | 8 | 1645 | XP_016873290.1 | |
AL512730.1 | 4 | 436 | CAC21663.1 | |
BC063280.1 | 22 | 2544 | AAH63280.1 | |
X80878.1 | 20 | 2602 | CAA56846.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |