Assay Details
Target Gene Details
Entrez Gene ID: | 51524 |
Gene Name: | transmembrane protein 138 |
Gene Aliases: |
HSPC196 |
Location: |
Chr.11:61362001-61376502 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM138 | NM_016464.4 | 5 | 1302 | NP_057548.1 |
NR_028473.1 | 6 | 1185 | ||
XM_006718585.3 | 4 | 1540 | XP_006718648.1 | |
XM_006718586.2 | 5 | 939 | XP_006718649.1 | |
XM_006718588.3 | 5 | 797 | XP_006718651.1 | |
XM_011545098.2 | XP_011543400.1 | |||
XM_011545099.1 | XP_011543401.1 | |||
XM_011545100.1 | XP_011543402.1 | |||
XM_017017917.1 | XP_016873406.1 | |||
AF151030.1 | 5 | 919 | AAF36116.1 | |
AF151032.1 | 5 | 916 | AAF36118.1 | |
AI768672.1 | ||||
AK027724.1 | 5 | 1451 | ||
AL832879.1 | 3 | 3456 | CAI46174.1 | |
BC005201.1 | 5 | 917 | AAH05201.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |