Assay Details
Target Gene Details
Entrez Gene ID: | 57010 |
Gene Name: | calcium binding protein 4 |
Gene Aliases: |
CRSD, CSNB2B |
Location: |
Chr.11:67452406-67461774 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABP4 | NM_001300896.1 | 1 | 61 | NP_001287825.1 |
XM_017018025.1 | 1 | 70 | XP_016873514.1 | |
AK311199.1 | 1 | 40 | ||
BC033167.1 | 1 | 61 | AAH33167.1 |
Target Gene Details
Entrez Gene ID: | 390212 |
Gene Name: | G protein-coupled receptor 152 |
Gene Aliases: |
PGR5 |
Location: |
Chr.11:67451301-67452729 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR152 | NM_206997.1 | 1 | 255 | NP_996880.1 |
AY255600.1 | AAO85112.1 | |||
AY569571.1 | 1 | 255 | AAS76893.1 | |
BC119779.1 | 1 | 255 | ||
BC122869.1 | 1 | 255 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | AIP RPS6KB2 CARNS1 CORO1B CABP4 PITPNM1 MIR6752 PTPRCAP GPR152 TMEM134 CDK2AP2 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | AIP CARNS1 RAD9A CABP4 PTPRCAP GPR152 CABP2 RPS6KB2 CORO1B PPP1CA PITPNM1 TBC1D10C MIR6752 TMEM134 CDK2AP2 |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | AIP CARNS1 RAD9A CABP4 PTPRCAP GPR152 CABP2 LOC100130987 CLCF1 RPS6KB2 CORO1B PPP1CA PITPNM1 TBC1D10C MIR6752 TMEM134 CDK2AP2 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | AIP CARNS1 CABP4 PTPRCAP GPR152 CABP2 RPS6KB2 CORO1B PITPNM1 TBC1D10C MIR6752 TMEM134 CDK2AP2 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | AIP CORO1B CABP4 PITPNM1 MIR6752 GPR152 TMEM134 CABP2 CDK2AP2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |