Assay Details
Target Gene Details
Entrez Gene ID: | 3321 |
Gene Name: | immunoglobulin superfamily member 3 |
Gene Aliases: |
EWI-3, LCDD, V8 |
Location: |
Chr.1:116574398-116667755 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGSF3 | NM_001007237.2 | 11 | 4218 | NP_001007238.1 |
NM_001542.3 | 11 | 4140 | NP_001533.2 | |
XM_005270793.2 | 12 | 4250 | XP_005270850.1 | |
XM_005270794.4 | 10 | 3488 | XP_005270851.1 | |
XM_006710593.3 | 11 | 3551 | XP_006710656.1 | |
XM_011541315.1 | 12 | 4302 | XP_011539617.1 | |
XM_011541316.2 | 6 | 3031 | XP_011539618.1 | |
AB007935.2 | 10 | 3550 | ||
AF031174.1 | 11 | 3618 | AAC72013.1 | |
BC110651.1 | 11 | 4221 | AAI10652.1 | |
BC130279.1 | 10 | 3891 | ||
BC144132.1 | 10 | 3891 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482103 | Chr.1:116574409 - 116667692 on Build GRCh38 | Gain | IGSF3 |
esv3578131 | Chr.1:116577298 - 116581814 on Build GRCh38 | Loss | IGSF3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |