Assay Details
Target Gene Details
Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP5SL | NM_001167867.1 | NP_001161339.1 | ||
NM_001167868.1 | NP_001161340.1 | |||
NM_001167869.1 | NP_001161341.1 | |||
NM_001167870.1 | NP_001161342.1 | |||
NM_001167871.1 | NP_001161343.1 | |||
NM_001320838.1 | NP_001307767.1 | |||
NM_001320840.1 | NP_001307769.1 | |||
NM_001320841.1 | NP_001307770.1 | |||
NM_018035.2 | NP_060505.2 | |||
NR_030765.1 | ||||
NR_135476.1 | ||||
NR_135477.1 | ||||
NR_135478.1 | ||||
XM_011527065.2 | XP_011525367.1 | |||
AK001103.1 | BAA91503.1 | |||
AK123631.1 | 3 | 1251 | ||
AK293132.1 | ||||
AK297698.1 | ||||
AK298197.1 | ||||
AK301940.1 | ||||
AK316313.1 | ||||
BC013323.1 | AAH13323.1 | |||
BQ072604.1 | ||||
DA540661.1 | ||||
DA631852.1 |