Assay Details
Target Gene Details
Entrez Gene ID: | 53635 |
Gene Name: | prostate tumor overexpressed 1 |
Gene Aliases: |
ACID2, PTOV-1 |
Location: |
Chr.19:49850726-49860744 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1 | NM_001305105.1 | NP_001292034.1 | ||
NM_001305108.1 | NP_001292037.1 | |||
NM_017432.4 | NP_059128.2 | |||
NR_130963.1 | ||||
XM_005258998.4 | XP_005259055.1 | |||
XM_006723244.3 | XP_006723307.1 | |||
XM_011527033.1 | XP_011525335.1 | |||
XM_011527034.2 | XP_011525336.1 | |||
XM_011527035.1 | XP_011525337.1 | |||
XM_017026879.1 | XP_016882368.1 | |||
XM_017026880.1 | XP_016882369.1 | |||
AF218026.1 | AAG17268.1 | |||
AJ617480.1 | CAE84582.1 | |||
AK057817.1 | ||||
AY358168.1 | AAQ88535.1 | |||
BC015172.1 | AAH15172.1 | |||
U79287.1 |
Target Gene Details
Entrez Gene ID: | 101928378 |
Gene Name: | PTOV1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.19:49856465-49859289 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1-AS2 | NR_110730.1 | |||
AL050131.1 | 1 | 456 | ||
CK903988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SLC17A7 MIR150 MIR4751 PRR12 TMEM143 RRAS PTOV1 SYNGR4 SNAR-A13 AKT1S1 ZNF473 SNAR-A3 SNAR-G1 SNAR-C5 MED25 MIR6799 CCDC114 SNORD35B SIGLEC16 CCDC155 SNAR-A8 TSKS GRIN2D SNAR-A10 GFY SNAR-A4 SNAR-B2 NUCB1 FCGRT BCAT2 PRRG2 SNAR-A12 NTN5 CPT1C LOC105447645 TRPM4 DHDH DKKL1 FLT3LG PTOV1-AS2 LOC101928295 CGB3 MIR5088 ALDH16A1 SLC6A16 SNAR-B1 CA11 CGB1 SNRNP70 SNAR-C2 SNAR-C4 PTH2 MIR6800 LHB C19orf73 PLA2G4C MIR6798 FUZ SIGLEC11 TBC1D17 RPS11 PRMT1 SEC1P CARD8-AS1 GYS1 GRWD1 SPACA4 SNAR-A7 RUVBL2 HRC SULT2B1 LMTK3 PPP1R15A TULP2 PLEKHA4 FLJ26850 FUT2 SNAR-A5 CGB2 PLA2G4C-AS1 RPL13A SNORD32A ATF5 CGB7 SCAF1 SNAR-A6 DBP MAMSTR IL4I1 CARD8 ZNF114 TEAD2 SNAR-C1 HSD17B14 PIH1D1 FTL SNAR-D PPFIA3 PTOV1-AS1 SNAR-A9 SNAR-G2 BSPH1 BAX ADM5 MIR4749 SNAR-A1 C19orf68 RASIP1 FUT1 RPL18 SNAR-C3 KDELR1 EMP3 IZUMO1 SPHK2 ELSPBP1 NTF4 LOC105372430 FGF21 BCL2L12 RCN3 VRK3 NOSIP FAM83E KCNA7 CD37 SNORD35A SNAR-A11 NUCB1-AS1 SNORD34 CABP5 MIR4324 CGB8 MIR4750 LIN7B SNAR-A14 CGB5 SNAR-A2 LOC101059948 LIG1 SNORD33 CYTH2 PNKP KCNJ14 NUP62 LOC100287477 IRF3 AP2A1 |
nsv526372 | Chr.19:49853907 - 49877601 on Build GRCh38 | Loss | MIR4749 PNKP TBC1D17 PTOV1 AKT1S1 PTOV1-AS2 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | IL4I1 MIR4750 FUZ MIR4751 PTOV1-AS1 TBC1D17 PTOV1 AKT1S1 MED25 MIR6799 MIR4749 PNKP NUP62 ATF5 TSKS MIR6800 AP2A1 PTOV1-AS2 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | MIR5088 MIR4751 PRR12 BCL2L12 RRAS NOSIP PTOV1 AKT1S1 MED25 MIR6799 ATF5 TSKS MIR6800 SCAF1 IL4I1 MIR4750 FUZ PTOV1-AS1 TBC1D17 PRMT1 ADM5 MIR4749 PNKP PRRG2 NUP62 CPT1C IRF3 AP2A1 PTOV1-AS2 |
dgv54n68 | Chr.19:49848512 - 50008788 on Build GRCh38 | Loss | IL4I1 MIR4750 MIR4751 VRK3 PTOV1-AS1 SIGLEC11 TBC1D17 PTOV1 AKT1S1 MIR4749 SIGLEC16 PNKP NUP62 ATF5 PTOV1-AS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |