Assay Details
Target Gene Details
Entrez Gene ID: | 57212 |
Gene Name: | TP73 antisense RNA 1 |
Gene Aliases: |
KIAA0495, PDAM |
Location: |
Chr.1:3735984-3747373 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP73-AS1 | NR_033708.1 | 1 | 1286 | |
NR_033709.1 | 1 | 1286 | ||
NR_033710.1 | 1 | 1286 | ||
NR_033711.1 | 1 | 1286 | ||
NR_033712.1 | 2 | 579 | ||
AB007964.1 | 1 | 1235 | ||
AL133648.1 | 1 | 539 | CAB63767.1 | |
BC093699.1 | 1 | 151 | AAH93699.1 | |
BC111984.1 | 1 | 151 | AAI11985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460061 | Chr.1:3680453 - 3755227 on Build GRCh38 | Loss | TP73 CCDC27 TP73-AS1 |
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss | MIR551A PRDM16 TPRG1L WRAP73 TP73 ARHGEF16 CCDC27 TP73-AS1 MEGF6 |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss | TPRG1L WRAP73 TP73 CCDC27 TP73-AS1 |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain | LINC01134 LRRC47 CEP104 TP73-AS1 MEGF6 LINC01346 DFFB TPRG1L WRAP73 TP73 C1orf174 SMIM1 CCDC27 |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain | AJAP1 LINC01134 LRRC47 ARHGEF16 CEP104 TP73-AS1 MEGF6 MIR551A LINC01346 DFFB TPRG1L WRAP73 TP73 C1orf174 LOC284661 SMIM1 CCDC27 |
nsv830425 | Chr.1:3740889 - 3841825 on Build GRCh38 | Gain | LRRC47 SMIM1 CEP104 CCDC27 TP73-AS1 |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion | PRDM16 LRRC47 LINC00982 ARHGEF16 TP73-AS1 MEGF6 MIR551A LOC105378604 TPRG1L WRAP73 MIR4251 TP73 SMIM1 CCDC27 |
nsv830314 | Chr.1:3660776 - 3804275 on Build GRCh38 | Gain+Loss | LRRC47 TP73 SMIM1 CCDC27 TP73-AS1 |
dgv13n100 | Chr.1:3730448 - 3791316 on Build GRCh38 | Gain | LRRC47 TP73 SMIM1 CCDC27 TP73-AS1 |