Assay Details
Target Gene Details
Entrez Gene ID: | 64333 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
10C, RGL1 |
Location: |
Chr.12:57472255-57488814 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain | DTX3 INHBE KIF5A DDIT3 GLI1 MBD6 DCTN2 MIR6758 MARS INHBC ARHGAP9 PIP4K2C MIR616 |
nsv819597 | Chr.12:57474778 - 57476427 on Build GRCh38 | Gain | ARHGAP9 |
nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss | DTX3 KIF5A DDIT3 GLI1 MBD6 DCTN2 MIR6758 ARHGEF25 MARS LOC101927583 ARHGAP9 PIP4K2C MIR616 |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss | INHBE MARS INHBC DDIT3 ARHGAP9 GLI1 MBD6 MIR6758 MIR616 |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | DTX3 KIF5A SLC39A5 TIMELESS IL23A GLI1 ANKRD52 DCTN2 ATP5B ARHGEF25 TAC3 MIR1228 R3HDM2 NXPH4 BAZ2A NABP2 PTGES3 RBMS2 RNF41 DDIT3 OS9 ZBTB39 PAN2 NAB2 MIR6758 MIP SDR9C7 GLS2 LRP1-AS PIP4K2C HSD17B6 SNORD59A GPR182 INHBE LRP1 NDUFA4L2 MBD6 STAC3 HBCBP NACA SNORA105C SHMT2 MARS NEMP1 SLC26A10 SPRYD4 CNPY2 MIR616 STAT6 RDH16 STAT2 SNORD59B INHBC LOC101927583 MYO1A CS ARHGAP9 COQ10A PRIM1 B4GALNT1 APOF |
nsv559035 | Chr.12:57467189 - 57476865 on Build GRCh38 | Loss | ARHGAP9 GLI1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |