Assay Details
Target Gene Details
Entrez Gene ID: | 7163 |
Gene Name: | tumor protein D52 |
Gene Aliases: |
D52, N8L, PC-1, PrLZ, hD52 |
Location: |
Chr.8:80034868-80171659 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPD52 | NM_001025252.2 | 6 | 2622 | NP_001020423.1 |
NM_001025253.2 | 8 | 2409 | NP_001020424.1 | |
NM_001287140.1 | 8 | 2691 | NP_001274069.1 | |
NM_001287142.1 | 7 | 2664 | NP_001274071.1 | |
NM_001287143.1 | 7 | 2649 | NP_001274072.1 | |
NM_001287144.1 | 6 | 2333 | NP_001274073.1 | |
NM_005079.3 | 6 | 2340 | NP_005070.1 | |
NR_105033.1 | 9 | 2761 | ||
NR_105034.1 | 5 | 2224 | ||
NR_105035.1 | 7 | 2410 | ||
NR_105036.1 | 8 | 2354 | ||
NR_105037.1 | 7 | 2355 | ||
AF202897.1 | AAO22156.1 | |||
AK057075.1 | ||||
BC018117.1 | AAH18117.1 | |||
BX640835.1 | CAE45908.1 | |||
DA079836.1 | 1 | 122 | ||
GQ499324.1 | ||||
GQ499325.1 | ||||
GQ499326.1 | ||||
GQ499327.1 | ||||
GQ499328.1 | ||||
S82081.1 | AAB36475.1 | |||
U18914.1 | 6 | 2215 | AAC50183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3617769 | Chr.8:80034764 - 80036567 on Build GRCh38 | Loss | TPD52 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |