Assay Details
Target Gene Details
Entrez Gene ID: | 3716 |
Gene Name: | Janus kinase 1 |
Gene Aliases: |
JAK1A, JAK1B, JTK3 |
Location: |
Chr.1:64833223-65067746 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JAK1 | NM_001320923.1 | NP_001307852.1 | ||
NM_001321852.1 | NP_001308781.1 | |||
NM_001321853.1 | NP_001308782.1 | |||
NM_001321854.1 | NP_001308783.1 | |||
NM_001321855.1 | NP_001308784.1 | |||
NM_001321856.1 | NP_001308785.1 | |||
NM_001321857.1 | NP_001308786.1 | |||
NM_002227.3 | NP_002218.2 | |||
XM_017001212.1 | XP_016856701.1 | |||
AB209057.1 | BAD92294.1 | |||
AB219242.1 | BAE02826.1 | |||
AK314257.1 | ||||
BC132729.1 | ||||
BX647675.1 | ||||
HQ628631.1 | ||||
M64174.1 | AAA36527.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |