Assay Details
Target Gene Details
Entrez Gene ID: | 9049 |
Gene Name: | aryl hydrocarbon receptor interacting protein |
Gene Aliases: |
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2 |
Location: |
Chr.11:67483029-67491108 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AIP | NM_001302959.1 | 2 | 307 | NP_001289888.1 |
NM_001302960.1 | 2 | 341 | NP_001289889.1 | |
NM_003977.3 | 2 | 341 | NP_003968.3 | |
BC104797.1 | 2 | 243 | AAI04798.1 | |
BC104827.1 | 2 | 243 | AAI04828.1 | |
BG821308.1 | 2 | 336 | ||
BQ054206.1 | 2 | 307 | ||
BQ420398.1 | 1 | 67 | ||
HY065637.1 | 2 | 341 | ||
U31913.1 | 2 | 220 | AAB39923.1 | |
U78521.1 | 2 | 328 | AAB59004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | CABP4 PITPNM1 RPS6KB2 TMEM134 CARNS1 PTPRCAP CDK2AP2 AIP MIR6752 CORO1B GPR152 |
esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication | PITPNM1 TMEM134 DOC2GP NDUFV1 C11orf72 ACY3 NUDT8 ALDH3B2 CDK2AP2 GSTP1 FAM86C2P CABP2 AIP MIR6752 TBX10 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | PITPNM1 RPS6KB2 TMEM134 CARNS1 PTPRCAP TBC1D10C RAD9A CABP4 CDK2AP2 PPP1CA CABP2 AIP MIR6752 CORO1B GPR152 |
nsv368 | Chr.11:67479842 - 67509866 on Build GRCh38 | Insertion | PITPNM1 CDK2AP2 AIP MIR6752 |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | PITPNM1 RPS6KB2 TMEM134 CARNS1 PTPRCAP TBC1D10C RAD9A CABP4 CDK2AP2 PPP1CA CABP2 AIP MIR6752 CORO1B CLCF1 LOC100130987 GPR152 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | PITPNM1 RPS6KB2 TMEM134 CARNS1 PTPRCAP TBC1D10C CABP4 CDK2AP2 CABP2 AIP MIR6752 CORO1B GPR152 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | CABP4 PITPNM1 TMEM134 CDK2AP2 CABP2 AIP MIR6752 CORO1B GPR152 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |