Assay Details
Target Gene Details
Entrez Gene ID: | 8996 |
Gene Name: | nucleolar protein 3 |
Gene Aliases: |
ARC, FCM, MYP, NOP, NOP30 |
Location: |
Chr.16:67170497-67175737 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOL3 | NM_001185057.2 | 4 | 891 | NP_001171986.1 |
NM_001276307.1 | 6 | 925 | NP_001263236.1 | |
NM_001276309.1 | 4 | 785 | NP_001263238.1 | |
NM_001276311.1 | 4 | 777 | NP_001263240.1 | |
NM_001276312.1 | 6 | 1276 | NP_001263241.1 | |
NM_001276319.1 | 6 | 1276 | NP_001263248.1 | |
NM_003946.6 | 4 | 901 | NP_003937.1 | |
XM_005256217.2 | 4 | 1528 | XP_005256274.1 | |
XM_005256219.3 | 4 | 1414 | XP_005256276.1 | |
XM_011523424.2 | 5 | 1373 | XP_011521726.1 | |
XM_017023843.1 | 4 | 1538 | XP_016879332.1 | |
AF043244.1 | 4 | 818 | AAC34993.1 | |
AF064599.1 | 4 | 742 | AAC18593.1 | |
AF064600.1 | 4 | 776 | AAC18594.1 | |
AK294145.1 | 6 | 1241 | ||
BC012798.2 | 4 | 787 | AAH12798.1 | |
BP215200.1 | ||||
BQ685512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)