Assay Details
Target Gene Details
Entrez Gene ID: | 377841 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492 |
Location: |
Chr.9:137434364-137441710 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENTPD8 | NM_001033113.1 | 10 | 1577 | NP_001028285.1 |
NM_198585.2 | 9 | 1466 | NP_940987.2 | |
XM_011518665.2 | 11 | 1883 | XP_011516967.1 | |
XM_011518666.2 | 10 | 1782 | XP_011516968.1 | |
XM_011518668.1 | 8 | 1238 | XP_011516970.1 | |
AY359088.1 | 9 | 1466 | AAQ89446.1 | |
AY430414.1 | 9 | 1410 | AAR04374.1 | |
AY903953.1 | 7 | 1640 | AAW83515.1 | |
AY903954.1 | 9 | 1393 | AAW83516.1 | |
BC141810.1 | 9 | 1367 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616021 | Chr.9:137434689 - 137452821 on Build GRCh38 | Loss | ENTPD8 MIR7114 NSMF |
nsv1049808 | Chr.9:137356366 - 137438026 on Build GRCh38 | Gain | ENTPD8 NOXA1 EXD3 |
nsv951208 | Chr.9:137379349 - 137467948 on Build GRCh38 | Deletion | ENTPD8 MIR7114 NOXA1 PNPLA7 NSMF EXD3 |
nsv6771 | Chr.9:137412670 - 137444666 on Build GRCh38 | Insertion | ENTPD8 NOXA1 EXD3 |
esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss | ENTPD8 NOXA1 EXD3 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | LCN12 TPRN MIR7114 MIR4292 SAPCD2 FUT7 LCNL1 TUBB4B TMEM203 TRAF2 LOC101930307 NOXA1 C8G MAN1B1 ENTPD2 FBXW5 CCDC183-AS1 RNF224 PNPLA7 SSNA1 LRRC26 EDF1 RABL6 ABCA2 MIR4479 CCDC183 C9orf139 RNF208 C9orf173-AS1 MAMDC4 NELFB UAP1L1 CYSRT1 C9orf172 NSMF STPG3 SLC34A3 CLIC3 PTGDS ENTPD8 TOR4A GRIN1 PHPT1 FAM166A MIR3621 TMEM210 NPDC1 EXD3 ANAPC2 DPP7 NDOR1 C9orf142 MAN1B1-AS1 NRARP PRR31 |
esv3622034 | Chr.9:137433077 - 137439026 on Build GRCh38 | Gain | ENTPD8 NOXA1 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | RNF224 ENTPD8 TPRN TOR4A TUBB4B GRIN1 RNF208 TMEM203 SSNA1 C9orf173-AS1 LRRC26 FAM166A NELFB MIR3621 TMEM210 EXD3 ANAPC2 CYSRT1 NDOR1 NOXA1 STPG3 SLC34A3 NRARP |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |