Assay Details
Target Gene Details
Entrez Gene ID: | 60343 |
Gene Name: | family with sequence similarity 3 member A |
Gene Aliases: |
2.19, DLD, DXS560S, XAP-7 |
Location: |
Chr.X:154506159-154516249 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | SLC10A3 G6PD MIR6858 CTAG2 TEX28 OPN1LW BRCC3 EMD MIR664B SNORA70 TMLHE CLIC2 FAM223A RAB39B MPP1 H2AFB1 IKBKG FAM50A F8A1 LOC107987332 SNORA56 MTCP1 ATP6AP1 PLXNA3 SMIM9 SNORA36A MECP2 CTAG1B F8 OPN1MW2 SPRY3 FLNA FAM3A IRAK1 F8A3 H2AFB2 F8A2 H2AFB3 GAB3 MIR1184-2 RPL10 FAM223B OPN1MW3 MIR3202-1 HCFC1 MIR3202-2 OPN1MW LOC101927830 DKC1 HCFC1-AS1 MIR1184-3 FUNDC2 MIR718 GDI1 LOC105373386 DNASE1L1 CMC4 TMEM187 TMLHE-AS1 MIR1184-1 VBP1 TKTL1 LAGE3 TAZ CTAG1A UBL4A CH17-340M24.3 |
nsv519042 | Chr.X:154082978 - 154601444 on Build GRCh38 | Gain | SLC10A3 G6PD MIR6858 RPL10 TEX28 OPN1LW OPN1MW3 EMD SNORA70 FAM223A IKBKG OPN1MW FAM50A LOC107987332 ATP6AP1 PLXNA3 GDI1 LOC105373386 DNASE1L1 MECP2 OPN1MW2 FLNA TKTL1 LAGE3 TAZ FAM3A CTAG1A UBL4A CH17-340M24.3 |
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | SLC10A3 G6PD MIR6858 TEX28 OPN1LW EMD SNORA70 PNCK LOC107983954 FAM223A TREX2 IKBKG FAM50A LOC107987332 HAUS7 ARHGAP4 ATP6AP1 PLXNA3 LOC105373381 ABCD1 MECP2 RENBP LCA10 OPN1MW2 BCAP31 PLXNB3 FLNA MAGEA1 ZNF275 FAM3A IRAK1 BGN LOC105373377 PDZD4 NAA10 RPL10 OPN1MW3 IDH3G MIR3202-1 L1CAM HCFC1 MIR3202-2 OPN1MW HCFC1-AS1 LOC105373383 DUSP9 PNMA6A MIR718 ZFP92 GDI1 LOC105373386 ATP2B3 DNASE1L1 TMEM187 SRPK3 SLC6A8 AVPR2 TKTL1 LAGE3 FAM58A TAZ CTAG1A UBL4A SSR4 LOC105373378 CH17-340M24.3 |
nsv508816 | Chr.X:154492171 - 154591783 on Build GRCh38 | Insertion | G6PD IKBKG FAM3A CTAG1A FAM223A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |