Assay Details
Target Gene Details
Entrez Gene ID: | 23583 |
Gene Name: | single-strand-selective monofunctional uracil-DNA glycosylase 1 |
Gene Aliases: |
FDG, HMUDG, UNG3 |
Location: |
Chr.12:54171815-54188994 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509467 | Chr.12:54173617 - 54255598 on Build GRCh38 | Insertion | MIR3198-2 CBX5 SMUG1 |
nsv832419 | Chr.12:54064043 - 54259156 on Build GRCh38 | Loss | FLJ12825 MIR3198-2 CBX5 SMUG1 LOC100240735 LOC100240734 LOC400043 |
nsv832420 | Chr.12:54137272 - 54296797 on Build GRCh38 | Loss | MIR3198-2 HNRNPA1 CBX5 SMUG1 NFE2 |
nsv832418 | Chr.12:54016556 - 54186660 on Build GRCh38 | Loss | FLJ12825 HOXC4 SMUG1 HOXC6 HOXC5 LOC100240735 LOC100240734 MIR615 LOC400043 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |