Assay Details
Target Gene Details
Entrez Gene ID: | 4798 |
Gene Name: | nuclear factor related to kappaB binding protein |
Gene Aliases: |
INO80G |
Location: |
Chr.11:129863773-129895596 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFRKB | NM_001143835.1 | NP_001137307.1 | ||
NM_006165.3 | NP_006156.2 | |||
XM_005271567.3 | XP_005271624.1 | |||
XM_005271568.3 | XP_005271625.1 | |||
XM_011542850.2 | XP_011541152.1 | |||
XM_011542851.2 | XP_011541153.1 | |||
XM_011542852.2 | XP_011541154.1 | |||
XM_017017793.1 | XP_016873282.1 | |||
XM_017017794.1 | XP_016873283.1 | |||
XM_017017795.1 | XP_016873284.1 | |||
XM_017017796.1 | XP_016873285.1 | |||
XM_017017797.1 | XP_016873286.1 | |||
XM_017017798.1 | XP_016873287.1 | |||
XM_017017799.1 | XP_016873288.1 | |||
XM_017017800.1 | XP_016873289.1 | |||
XM_017017801.1 | XP_016873290.1 | |||
AL512730.1 | CAC21663.1 | |||
BC063280.1 | AAH63280.1 | |||
X80878.1 | CAA56846.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |