Assay Details
Target Gene Details
Entrez Gene ID: | 5650 |
Gene Name: | kallikrein related peptidase 7 |
Gene Aliases: |
PRSS6, SCCE, hK7 |
Location: |
Chr.19:50976479-50984064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLK7 | NM_001207053.1 | NP_001193982.1 | ||
NM_001243126.1 | NP_001230055.1 | |||
NM_005046.3 | NP_005037.1 | |||
NM_139277.2 | NP_644806.1 | |||
AA101043.1 | ||||
AF411214.1 | AAN03662.1 | |||
AF411215.1 | AAN03663.1 | |||
AK289660.1 | ||||
AK295313.1 | ||||
AY601109.1 | ||||
AY646152.1 | AAT66047.1 | |||
BC032005.1 | AAH32005.1 | |||
DQ018784.1 | ||||
KF963189.1 | ||||
KF963190.1 | ||||
KF963191.1 | ||||
KF963192.1 | ||||
KF963193.1 | ||||
L33404.1 | AAC37551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv513532 | Chr.19:50980352 - 50981443 on Build GRCh38 | Insertion | KLK7 |
nsv1064207 | Chr.19:50971269 - 50994022 on Build GRCh38 | Loss | KLK7 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |