Assay Details
Target Gene Details
Entrez Gene ID: | 5368 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ |
Location: |
Chr.8:28316979-28343355 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PNOC | NM_001284244.1 | 2 | 488 | NP_001271173.1 |
NM_006228.4 | 3 | 681 | NP_006219.1 | |
XM_005273532.2 | XP_005273589.1 | |||
XM_011544559.2 | XP_011542861.1 | |||
XM_017013578.1 | XP_016869067.1 | |||
AK289633.1 | 3 | 544 | ||
AK301456.1 | 2 | 488 | ||
AY335948.1 | AAP94601.1 | |||
BC034758.1 | 3 | 526 | AAH34758.1 | |
BF976583.1 | 2 | 468 | ||
BT019334.1 | AAV38141.1 | |||
CA412176.1 | ||||
CR541940.1 | CAG46738.1 | |||
CR541965.1 | CAG46763.1 | |||
U48263.1 | 3 | 684 | AAC50651.1 | |
X97370.1 | 2 | 496 | CAA66040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv456n21 | Chr.8:28321496 - 28467894 on Build GRCh38 | Gain | ZNF395 PNOC FBXO16 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |