Assay Details
Target Gene Details
Entrez Gene ID: | 53832 |
Gene Name: | interleukin 20 receptor subunit alpha |
Gene Aliases: |
CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA |
Location: |
Chr.6:136999971-137045366 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL20RA | NM_001278722.1 | 7 | 1224 | NP_001265651.1 |
NM_001278723.1 | 6 | 1364 | NP_001265652.1 | |
NM_001278724.1 | 6 | 1298 | NP_001265653.1 | |
NM_014432.3 | 7 | 1543 | NP_055247.3 | |
XM_006715506.2 | 6 | 940 | XP_006715569.1 | |
XM_011535904.2 | 6 | 1051 | XP_011534206.1 | |
XM_017010954.1 | 7 | 1732 | XP_016866443.1 | |
XM_017010955.1 | 7 | 1192 | XP_016866444.1 | |
AF184971.1 | 7 | 1327 | AAF01320.1 | |
AJ420490.1 | 3 | 518 | ||
AK098312.1 | 3 | 1133 | ||
AK297121.1 | 7 | 1186 | ||
AK315293.1 | 7 | 1331 | ||
AY358883.1 | 6 | 878 | AAQ89242.1 | |
BC113574.1 | 7 | 1421 | ||
BC113602.1 | 7 | 1421 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |