Assay Details
Target Gene Details
Entrez Gene ID: | 9477 |
Gene Name: | mediator complex subunit 20 |
Gene Aliases: |
PRO0213, TRFP |
Location: |
Chr.6:41905353-41921147 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED20 | NM_001305455.1 | 3 | 461 | NP_001292384.1 |
NM_001305456.1 | 5 | 733 | NP_001292385.1 | |
NM_001305457.1 | 4 | 540 | NP_001292386.1 | |
NM_004275.4 | 4 | 616 | NP_004266.2 | |
NR_131160.1 | 3 | 362 | ||
AF097725.1 | 4 | 565 | AAD16169.1 | |
AJ420544.1 | 3 | 419 | ||
AK023092.1 | 4 | 560 | BAB14399.1 | |
AK097385.1 | 5 | 733 | ||
AK293412.1 | 4 | 512 | ||
AK300573.1 | 3 | 453 | ||
AK307070.1 | 3 | 354 | ||
AL050196.1 | 2 | 131 | CAB43314.1 | |
BC012618.1 | 4 | 612 | AAH12618.1 | |
BC019866.1 | 4 | 593 | ||
BC032552.1 | 4 | 578 | AAH32552.1 | |
BC040950.1 | 4 | 552 | AAH40950.1 | |
BU597139.1 | 4 | 611 | ||
DA909832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5278 | Chr.6:41906650 - 41908185 on Build GRCh38 | Insertion | MED20 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |