Assay Details
Target Gene Details
Entrez Gene ID: | 100500847 |
Gene Name: | microRNA 3615 |
Gene Aliases: |
mir-3615 |
Location: |
Chr.17:74748613-74748699 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIR3615 | NR_037409.1 |
Target Gene Details
Entrez Gene ID: | 9368 |
Gene Name: | SLC9A3 regulator 1 |
Gene Aliases: |
EBP50, NHERF, NHERF-1, NHERF1, NPHLOP2 |
Location: |
Chr.17:74748612-74769360 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC9A3R1 | NM_004252.4 | 1 | 138 | NP_004243.1 |
AF015926.1 | 1 | 115 | AAC52084.1 | |
AF036241.1 | 1 | 73 | AAC04572.1 | |
AK026581.1 | 1 | 121 | ||
AK094467.1 | 1 | 122 | ||
AK128474.1 | 1 | 122 | ||
AK312498.1 | 1 | 154 | ||
BC001443.2 | 1 | 91 | AAH01443.1 | |
BC003361.1 | 1 | 108 | AAH03361.1 | |
BC011777.1 | 1 | 107 | AAH11777.1 | |
BC049220.1 | 1 | 114 | AAH49220.1 | |
BC053350.1 | 1 | 98 | AAH53350.1 | |
BP347679.1 | 1 | 126 | ||
CX761192.1 | 1 | 138 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |