Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOPEY1 | XM_011535619.2 | 41 | 8390 | XP_011533921.1 |
XM_017010559.1 | 42 | 8420 | XP_016866048.1 | |
XM_017010560.1 | 41 | 8348 | XP_016866049.1 | |
XM_017010561.1 | 42 | 8372 | XP_016866050.1 | |
XM_017010562.1 | 41 | 8321 | XP_016866051.1 | |
XM_017010563.1 | 40 | 9060 | XP_016866052.1 | |
XM_017010564.1 | 41 | 8339 | XP_016866053.1 | |
XM_017010565.1 | 40 | 9054 | XP_016866054.1 | |
XM_017010566.1 | 41 | 8333 | XP_016866055.1 | |
XM_017010567.1 | 39 | 8982 | XP_016866056.1 | |
XM_017010568.1 | 39 | 9051 | XP_016866057.1 | |
XM_017010569.1 | 38 | 8968 | XP_016866058.1 | |
XM_017010570.1 | 41 | 8321 | XP_016866059.1 | |
XM_017010571.1 | 41 | 8315 | XP_016866060.1 | |
XM_017010572.1 | 41 | 8270 | XP_016866061.1 |
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PGM3 | NM_001199917.1 | 13 | 1748 | NP_001186846.1 |
NM_001199918.1 | 11 | 1406 | NP_001186847.1 | |
NM_001199919.1 | 12 | 1612 | NP_001186848.1 | |
NM_015599.2 | 12 | 1612 | NP_056414.1 | |
XM_017010935.1 | 11 | 1379 | XP_016866424.1 | |
XM_017010936.1 | 12 | 1611 | XP_016866425.1 | |
XM_017010937.1 | 10 | 1256 | XP_016866426.1 | |
XM_017010938.1 | 11 | 1477 | XP_016866427.1 | |
AB032081.1 | 12 | 1524 | BAB00613.1 | |
AF102265.1 | 12 | 1528 | AAC72409.1 | |
AF180371.1 | 12 | 1535 | AAD55097.1 | |
AK001365.1 | 11 | 1347 | ||
AK023432.1 | 12 | 1557 | ||
AK023709.1 | 2 | 238 | BAB14652.1 | |
AK225363.1 | 12 | 1550 | ||
AK301867.1 | 13 | 1726 | ||
AK314512.1 | 12 | 1545 | ||
AL117443.1 | 3 | 323 | CAB55928.1 | |
BC001258.1 | 12 | 1571 | AAH01258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain | RWDD2A UBE3D PGM3 DOPEY1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |