Assay Details
Target Gene Details
Entrez Gene ID: | 8681 |
Gene Name: | JMJD7-PLA2G4B readthrough |
Gene Aliases: |
HsT16992, cPLA2-beta |
Location: |
Chr.15:41828085-41848148 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JMJD7-PLA2G4B | NM_001198588.1 | NP_001185517.1 | ||
NM_005090.3 | NP_005081.1 | |||
AB209150.1 | BAD92387.1 | |||
AF065215.1 | AAC78836.1 | |||
AF065216.1 | ||||
AF121908.1 | AAD32135.1 | |||
BI768993.1 | ||||
DQ523799.1 | ||||
DQ523800.1 |
Target Gene Details
Entrez Gene ID: | 100137049 |
Gene Name: | phospholipase A2 group IVB |
Gene Aliases: |
HsT16992, cPLA2-beta |
Location: |
Chr.15:41838813-41848148 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G4B | NM_001114633.1 | NP_001108105.1 | ||
AK124489.1 | ||||
AK299419.1 | ||||
BF372565.1 | ||||
BX648318.1 | ||||
CA488226.1 | ||||
CU447449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9244 | Chr.15:41818514 - 41916607 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1050892 | Chr.15:41836657 - 41894284 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1506 | Chr.15:41824733 - 41857173 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() |
nsv952613 | Chr.15:41816803 - 41884902 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv517631 | Chr.15:41804728 - 41895294 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv2582n100 | Chr.15:41791376 - 41900562 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)