Assay Details
Target Gene Details
Entrez Gene ID: | 6150 |
Gene Name: | mitochondrial ribosomal protein L23 |
Gene Aliases: |
L23MRP, RPL23, RPL23L |
Location: |
Chr.11:1947272-2013176 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL23 | NM_021134.3 | NP_066957.3 | ||
XM_011520276.2 | XP_011518578.1 | |||
AK128774.1 | ||||
AK313805.1 | ||||
BC027710.2 | AAH27710.1 | |||
U26596.1 | AAC50415.1 | |||
Z49254.1 | CAA89220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2743985 | Chr.11:1950054 - 1951240 on Build GRCh38 | Deletion |
![]() |
nsv1047474 | Chr.11:1803515 - 2043381 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1016n100 | Chr.11:1946041 - 2043381 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv825703 | Chr.11:1940540 - 1957159 on Build GRCh38 | Gain |
![]() ![]() |
nsv553064 | Chr.11:1943082 - 2001669 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv516622 | Chr.11:1927731 - 1993479 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv469926 | Chr.11:1813024 - 2036416 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3579243 | Chr.11:1939895 - 1955789 on Build GRCh38 | Loss |
![]() ![]() |
esv2041292 | Chr.11:1950430 - 1950873 on Build GRCh38 | Deletion |
![]() |
esv29980 | Chr.11:1820597 - 2097889 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv522320 | Chr.11:1835882 - 1996938 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2743987 | Chr.11:1950396 - 1950985 on Build GRCh38 | Deletion |
![]() |
nsv951283 | Chr.11:1913571 - 2040070 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)