Assay Details
Target Gene Details
Entrez Gene ID: | 11274 |
Gene Name: | ubiquitin specific peptidase 18 |
Gene Aliases: |
ISG43, UBP43 |
Location: |
Chr.22:18149959-18177397 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP18 | NM_017414.3 | 2 | 354 | NP_059110.2 |
XM_006724074.2 | 2 | 386 | XP_006724137.1 | |
AF176642.2 | AAD49967.1 | |||
AI222587.1 | 1 | 64 | ||
AJ243526.1 | 2 | 219 | CAB76398.1 | |
AK313385.1 | 2 | 175 | ||
AL136690.1 | 2 | 175 | CAB66625.1 | |
BC014896.1 | 2 | 160 | AAH14896.1 | |
BI752849.1 | 2 | 250 | ||
BT006835.1 | AAP35481.1 | |||
CR457216.1 | CAG33497.1 | |||
CT841507.1 | 1 | 82 | CAJ86437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv173e55 | Chr.22:18157533 - 19070277 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv516997 | Chr.22:18143870 - 19039100 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1055756 | Chr.22:18062621 - 18794021 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1058565 | Chr.22:18060494 - 18159378 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2751935 | Chr.22:18149609 - 19042823 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3893430 | Chr.22:18059545 - 18169093 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2758824 | Chr.22:18069029 - 19074200 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471594 | Chr.22:18076012 - 18882129 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469560 | Chr.22:18076011 - 18213970 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)