Assay Details
Target Gene Details
Entrez Gene ID: | 4582 |
Gene Name: | mucin 1, cell surface associated |
Gene Aliases: |
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM |
Location: |
Chr.1:155185824-155192915 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv1005636 | Chr.1:155187511 - 155193592 on Build GRCh38 | Insertion |
![]() |
nsv527682 | Chr.1:155177242 - 155227671 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv470741 | Chr.1:155142784 - 155260340 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1010146 | Chr.1:155146620 - 155231273 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv2988 | Chr.1:155182130 - 155219701 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv547961 | Chr.1:155109822 - 155225189 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)