Assay Details
Target Gene Details
Entrez Gene ID: | 1496 |
Gene Name: | catenin alpha 2 |
Gene Aliases: |
CAP-R, CAPR, CT114, CTNR |
Location: |
Chr.2:79512934-80648868 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTNNA2 | NM_001164883.1 | NP_001158355.1 | ||
NM_001282597.2 | NP_001269526.1 | |||
NM_001282598.1 | NP_001269527.1 | |||
NM_004389.3 | NP_004380.2 | |||
XM_011532555.2 | XP_011530857.1 | |||
XM_011532556.2 | XP_011530858.1 | |||
XM_017003403.1 | XP_016858892.1 | |||
XM_017003404.1 | XP_016858893.1 | |||
XM_017003405.1 | XP_016858894.1 | |||
AK127226.1 | ||||
AK295181.1 | ||||
AK303035.1 | ||||
BC040458.1 | AAH40458.1 | |||
BC052996.1 | AAH52996.1 | |||
M94151.2 | AAA58407.2 |
Target Gene Details
Entrez Gene ID: | 347730 |
Gene Name: | leucine rich repeat transmembrane neuronal 1 |
Gene Aliases: |
- |
Location: |
Chr.2:80301878-80304738 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRTM1 | NM_178839.4 | NP_849161.2 | ||
XM_017003986.1 | 1 | 377 | XP_016859475.1 | |
XM_017003987.1 | 1 | 377 | XP_016859476.1 | |
AK056644.1 | 1 | 32 | BAB71240.1 | |
AK225779.1 | 1 | 388 | ||
AK290512.1 | 1 | 251 | ||
AY358310.1 | 1 | 450 | AAQ88677.1 | |
BI756350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759069 | Chr.2:80174192 - 80615336 on Build GRCh38 | Loss | LRRTM1 CTNNA2 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |