Assay Details
Target Gene Details
Entrez Gene ID: | 64403 |
Gene Name: | cadherin 24 |
Gene Aliases: |
CDH11L |
Location: |
Chr.14:23047061-23057619 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH24 | NM_022478.3 | 3 | 545 | NP_071923.2 |
NM_144985.3 | 3 | 545 | NP_659422.2 | |
XM_011537089.1 | 3 | 626 | XP_011535391.1 | |
AK057922.1 | 3 | 481 | BAB71613.1 | |
AK289576.1 | 3 | 545 | ||
AK307746.1 | 3 | 527 | ||
AY260900.1 | 2 | 378 | AAP20590.1 | |
AY260901.1 | 2 | 378 | AAP20591.1 | |
AY358199.1 | 3 | 501 | AAQ88566.1 | |
BC110867.1 | 3 | 516 | AAI10868.1 | |
BX248750.1 | 3 | 527 | CAD66557.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |