Assay Details
Target Gene Details
Entrez Gene ID: | 27243 |
Gene Name: | charged multivesicular body protein 2A |
Gene Aliases: |
BC-2, BC2, CHMP2, VPS2, VPS2A |
Location: |
Chr.19:58551566-58555624 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHMP2A | NM_014453.3 | 4 | 550 | NP_055268.1 |
NM_198426.2 | 4 | 552 | NP_940818.1 | |
XM_005258746.2 | 4 | 1050 | XP_005258803.1 | |
XM_005258747.3 | 4 | 1173 | XP_005258804.1 | |
AF042384.1 | 4 | 518 | AAC00005.1 | |
AK311974.1 | 4 | 438 | ||
AY364248.1 | 3 | 389 | AAQ76807.1 | |
BC002502.2 | 4 | 522 | AAH02502.1 | |
BE856471.1 | 3 | 366 | ||
BG546479.1 | 4 | 546 | ||
BI457978.1 | 4 | 542 | ||
BP315111.1 | ||||
BT007298.1 | 3 | 389 | AAP35962.1 | |
BU147101.1 | 2 | 197 | ||
CN305066.1 | 4 | 553 | ||
CR457002.1 | 3 | 389 | CAG33283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057206 | Chr.19:58546716 - 58586475 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3687n100 | Chr.19:58473466 - 58586475 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv515731 | Chr.19:58320453 - 58576112 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv820066 | Chr.19:58551887 - 58552522 on Build GRCh38 | Loss |
![]() |
nsv952156 | Chr.19:58466134 - 58600733 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160670 | Chr.19:58433008 - 58586487 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv828657 | Chr.19:58483484 - 58585740 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3688n100 | Chr.19:58506055 - 58586475 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1057077 | Chr.19:58411953 - 58586475 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)