Assay Details
Target Gene Details
Entrez Gene ID: | 285203 |
Gene Name: | EGF domain specific O-linked N-acetylglucosamine transferase |
Gene Aliases: |
AER61, AOS4, C3orf64, EOGT1 |
Location: |
Chr.3:68975212-69013961 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EOGT | NM_001278689.1 | 18 | 2385 | NP_001265618.1 |
NM_173654.2 | 15 | 2133 | NP_775925.1 | |
NR_103826.1 | 16 | 2174 | ||
XM_005264743.2 | 18 | 2599 | XP_005264800.1 | |
XM_011533599.1 | 19 | 2653 | XP_011531901.1 | |
XM_011533600.1 | 19 | 2506 | XP_011531902.1 | |
XM_011533602.1 | 19 | 2418 | XP_011531904.1 | |
XM_011533603.1 | 19 | 2650 | XP_011531905.1 | |
XM_017006204.1 | 19 | 2503 | XP_016861693.1 | |
XM_017006205.1 | 18 | 2364 | XP_016861694.1 | |
XM_017006206.1 | 18 | 2260 | XP_016861695.1 | |
XM_017006207.1 | 11 | 1099 | XP_016861696.1 | |
XM_017006209.1 | 9 | 972 | XP_016861698.1 | |
AK023140.1 | ||||
AK091089.1 | 9 | 1363 | ||
AK123213.1 | 6 | 697 | ||
AK126187.1 | 14 | 1457 | BAC86479.1 | |
AK290356.1 | 14 | 1512 | ||
AK294101.1 | 10 | 1025 | ||
AK304102.1 | 16 | 1930 | ||
AL833467.1 | 4 | 539 | ||
BC060887.1 | 15 | 1862 | AAH60887.1 | |
BX640821.1 | 14 | 1506 | CAE45897.2 | |
BX648818.1 | 18 | 2160 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |