Assay Details
Target Gene Details
Entrez Gene ID: | 10367 |
Gene Name: | mitochondrial calcium uptake 1 |
Gene Aliases: |
CALC, CBARA1, EFHA3, MPXPS |
Location: |
Chr.10:72367326-72626191 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MICU1 | NM_001195518.1 | 12 | 1507 | NP_001182447.1 |
NM_001195519.1 | 7 | 857 | NP_001182448.1 | |
NM_006077.3 | 13 | 1513 | NP_006068.2 | |
XM_005269383.2 | 12 | 1429 | XP_005269440.1 | |
XM_005269386.2 | 8 | 970 | XP_005269443.1 | |
XM_011539119.1 | 13 | 1579 | XP_011537421.1 | |
AK022697.1 | 10 | 972 | BAB14187.1 | |
AK023318.1 | 12 | 1426 | ||
AK225298.1 | 12 | 1411 | ||
AK225695.1 | 12 | 1395 | ||
AK296086.1 | 7 | 824 | ||
AK298347.1 | 9 | 1033 | ||
AL117423.1 | 13 | 1404 | CAB55915.1 | |
BC004190.2 | 12 | 1419 | AAH04190.1 | |
BC004216.2 | 12 | 1419 | AAH04216.1 | |
BC016641.1 | 12 | 1382 | AAH16641.1 | |
Y17711.1 | 8 | 835 | CAA76830.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |