Assay Details
Target Gene Details
Entrez Gene ID: | 56605 |
Gene Name: | endoplasmic reticulum oxidoreductase 1 beta |
Gene Aliases: |
ERO1LB, Ero1beta |
Location: |
Chr.1:236215122-236282039 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERO1B | NM_019891.3 | 16 | 2301 | NP_063944.3 |
AK001865.1 | 1 | 215 | ||
BC032823.2 | 16 | 2152 | AAH32823.2 | |
BC044573.1 | 16 | 2247 | AAH44573.1 | |
BX648202.1 | 1 | 627 | ||
CR749523.1 | 1 | 488 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv590n100 | Chr.1:236139422 - 236267713 on Build GRCh38 | Gain | GPR137B ERO1B |
nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain | ACTN2 LGALS8 NID1 MT1HL1 LGALS8-AS1 EDARADD MTR HEATR1 GPR137B ERO1B |
nsv522991 | Chr.1:236215849 - 236250146 on Build GRCh38 | Loss | ERO1B |
nsv832969 | Chr.1:236111012 - 236311443 on Build GRCh38 | Gain | GPR137B ERO1B |
esv3578490 | Chr.1:236180905 - 236219922 on Build GRCh38 | Loss | GPR137B ERO1B |
nsv549394 | Chr.1:235876577 - 236629706 on Build GRCh38 | Gain | LYST LGALS8 NID1 LGALS8-AS1 EDARADD HEATR1 GPR137B ERO1B |
dgv156e212 | Chr.1:236206561 - 236219922 on Build GRCh38 | Loss | GPR137B ERO1B |
nsv1012436 | Chr.1:235860657 - 236648100 on Build GRCh38 | Loss | LYST LGALS8 NID1 LGALS8-AS1 EDARADD HEATR1 GPR137B ERO1B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |