Assay Details
Target Gene Details
Entrez Gene ID: | 53635 |
Gene Name: | prostate tumor overexpressed 1 |
Gene Aliases: |
ACID2, PTOV-1 |
Location: |
Chr.19:49850726-49860744 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1 | NM_001305105.1 | NP_001292034.1 | ||
NM_001305108.1 | NP_001292037.1 | |||
NM_017432.4 | NP_059128.2 | |||
NR_130963.1 | ||||
XM_005258998.4 | XP_005259055.1 | |||
XM_006723244.3 | XP_006723307.1 | |||
XM_011527033.1 | XP_011525335.1 | |||
XM_011527034.2 | XP_011525336.1 | |||
XM_011527035.1 | XP_011525337.1 | |||
XM_017026879.1 | XP_016882368.1 | |||
XM_017026880.1 | XP_016882369.1 | |||
AJ617480.1 | CAE84582.1 | |||
AK057817.1 | ||||
AK294366.1 | ||||
AY358168.1 | AAQ88535.1 | |||
U79287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | FCGRT PRMT1 SNAR-A7 ADM5 CA11 SNAR-A14 SNORD33 MIR4750 SCAF1 CGB5 SNAR-G1 PIH1D1 FLT3LG SNAR-A13 CCDC114 PRR12 SNORD34 BCAT2 IZUMO1 C19orf73 SPACA4 RRAS DBP SYNGR4 SNAR-C4 SNAR-B1 KCNJ14 HRC SNORD32A LIN7B FUZ ATF5 SNAR-C5 NUCB1-AS1 PTOV1-AS1 SNAR-C2 MED25 SULT2B1 C19orf68 SNAR-D NOSIP MIR6798 FAM83E LOC100287477 SNAR-C1 FTL SNORD35B LOC101059948 SNORD35A TRPM4 FGF21 SNAR-A8 FUT2 SNAR-A5 LMTK3 RPL18 SNAR-A4 RASIP1 MIR4751 CGB3 SNAR-A12 ELSPBP1 MIR4324 TSKS RPL13A LHB MIR6800 KCNA7 CGB7 FLJ26850 TMEM143 SIGLEC16 FUT1 DHDH IRF3 DKKL1 PPP1R15A SLC17A7 NTF4 LOC105372430 AKT1S1 AP2A1 PTOV1-AS2 GYS1 BCL2L12 SNAR-A11 PRRG2 CARD8-AS1 GRIN2D SEC1P SNAR-A10 SNRNP70 MIR150 CABP5 PLA2G4C-AS1 PTH2 IL4I1 CGB8 SIGLEC11 KDELR1 TBC1D17 CD37 MAMSTR SNAR-A9 CCDC155 HSD17B14 RCN3 MIR6799 GRWD1 CYTH2 RPS11 SLC6A16 SNAR-G2 SNAR-B2 CGB2 LIG1 ZNF114 RUVBL2 SNAR-A3 CGB1 PLEKHA4 SNAR-A2 LOC101928295 MIR5088 SNAR-A1 LOC105447645 GFY EMP3 SNAR-A6 ZNF473 NUP62 SNAR-C3 VRK3 PTOV1 PNKP PLA2G4C CPT1C TEAD2 NTN5 ALDH16A1 TULP2 MIR4749 NUCB1 SPHK2 CARD8 BSPH1 PPFIA3 BAX |
nsv526372 | Chr.19:49853907 - 49877601 on Build GRCh38 | Loss | TBC1D17 AKT1S1 MIR4749 PTOV1-AS2 PTOV1 PNKP |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | MIR6799 BCL2L12 PRR12 MIR6800 PRMT1 RRAS PTOV1 ADM5 IRF3 CPT1C FUZ PTOV1-AS1 AP2A1 MED25 MIR5088 SCAF1 MIR4749 PTOV1-AS2 TSKS |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | MIR6799 MIR6800 NUP62 IL4I1 PTOV1 PNKP FUZ ATF5 TBC1D17 PTOV1-AS1 AKT1S1 AP2A1 MIR4750 MED25 MIR4749 PTOV1-AS2 MIR4751 TSKS |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | MIR6799 BCL2L12 PRRG2 PRMT1 ADM5 MIR4750 MIR5088 SCAF1 MIR4751 TSKS PRR12 MIR6800 NUP62 RRAS IL4I1 PTOV1 PNKP IRF3 CPT1C FUZ ATF5 TBC1D17 PTOV1-AS1 AKT1S1 AP2A1 MED25 MIR4749 PTOV1-AS2 NOSIP |
dgv54n68 | Chr.19:49848512 - 50008788 on Build GRCh38 | Loss | NUP62 SIGLEC16 VRK3 IL4I1 PTOV1 PNKP SIGLEC11 ATF5 TBC1D17 PTOV1-AS1 AKT1S1 MIR4750 MIR4749 PTOV1-AS2 MIR4751 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |