Assay Details
Target Gene Details
Entrez Gene ID: | 84818 |
Gene Name: | interleukin 17 receptor C |
Gene Aliases: |
CANDF9, IL17-RL, IL17RL |
Location: |
Chr.3:9917074-9933627 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv589648 | Chr.3:9892921 - 10042251 on Build GRCh38 | Loss | PRRT3-AS1 LOC401052 EMC3 FANCD2 PRRT3 CIDECP JAGN1 IL17RE IL17RC EMC3-AS1 CRELD1 |
nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss | RPUSD3 CIDEC EMC3 PRRT3 TTLL3 ARPC4-TTLL3 IL17RC ARPC4 PRRT3-AS1 TADA3 LOC107986062 JAGN1 IL17RE CRELD1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |