Assay Details
Target Gene Details
Entrez Gene ID: | 10075 |
Gene Name: | HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase |
Gene Aliases: |
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1 |
Location: |
Chr.X:53532096-53686729 on Build GRCh38 |
Assay Gene Location: | Within Exon 86 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | GPR173 PHF8 SMC1A FAM120C MTRNR2L10 MIR4536-1 PAGE3 IQSEC2 SNORA109 FAM156A MAGEH1 GNL3L MIRLET7F2 MIR6857 KANTR TSR2 HSD17B10 LOC100421746 ALAS2 PAGE2 FGD1 TSPYL2 HUWE1 USP51 WNK3 APEX2 SNORA11 XAGE3 TRO MAGED2 PFKFB1 PAGE5 ITIH6 FOXR2 MIR6894 KDM5C MIR6895 FAM104B FAM156B RIBC1 MIR4536-2 MIR98 PAGE2B |
esv3576879 | Chr.X:53529972 - 53593214 on Build GRCh38 | Gain | MIRLET7F2 HUWE1 MIR98 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | GPR173 PHF8 SMC1A FAM120C MTRNR2L10 MIR4536-1 PAGE3 IQSEC2 SNORA109 FAM156A MAGEH1 GNL3L MIRLET7F2 MIR6857 KANTR TSR2 HSD17B10 LOC100421746 ALAS2 PAGE2 FGD1 TSPYL2 HUWE1 USP51 WNK3 APEX2 SNORA11 XAGE3 TRO MAGED2 PFKFB1 PAGE5 ITIH6 FOXR2 MIR6894 KDM5C MIR6895 FAM104B FAM156B RIBC1 MIR4536-2 MIR98 PAGE2B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |