Assay Details
Target Gene Details
Entrez Gene ID: | 283870 |
Gene Name: | BRICHOS domain containing 5 |
Gene Aliases: |
C16orf79 |
Location: |
Chr.16:2209253-2213037 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BRICD5 | NM_182563.3 | NP_872369.2 | ||
XM_005255263.4 | XP_005255320.1 | |||
AY597811.1 | AAT09004.1 | |||
BC039154.2 | AAH39154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571200 | Chr.16:2209190 - 2244889 on Build GRCh38 | Loss | DNASE1L2 PGP BRICD5 E4F1 MLST8 ECI1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | RPS2 RGS11 SYNGR3 TSC2 TPSB2 CACNA1H RNF151 ATP6V0C ABCA17P DDX11L10 MIR3177 SPSB3 HN1L ARHGDIG SNORA78 MIR6767 FAM234A PRSS27 NUBP2 HS3ST6 C1QTNF8 DNASE1L2 C16orf91 WDR90 CASKIN1 WDR24 PGP METRN NHLRC4 POLR3K NPW LOC100134368 NARFL CHTF18 NTN3 RPL23AP5 RAB40C PDPK1 MEIOB AXIN1 TPSAB1 MIR6768 LMF1-AS1 SSTR5-AS1 UNKL MRPL28 DECR2 LOC105371038 WFIKKN1 LOC105371049 SNRNP25 NTHL1 HBA1 CEMP1 CCDC154 GNG13 SNORD60 LOC105371047 SNHG9 HBQ1 NPRL3 LOC100287175 IFT140 NME4 RAB11FIP3 SLC9A3R2 FBXL16 TMEM204 MPG SOX8 MIR662 EME2 PTX4 MRPS34 MIR4717 NDUFB10 MIR3677 MCRIP2 MIR940 MIR4516 CRAMP1 UBE2I CCNF METTL26 MIR5587 GFER LOC105371045 BRICD5 TPSD1 MSLN STUB1 MIR3176 RAB26 RNPS1 HAGHL GNPTG TPSG1 TRAF7 HBZ LINC00235 NME3 PRR35 PRR25 PDIA2 HBM LOC105371184 TELO2 FLJ42627 CCDC78 JMJD8 WASIR2 MAPK8IP3 MLST8 AMDHD2 TBL3 PIGQ ERVK13-1 IGFALS LMF1 LOC107984876 LOC729652 PKD1 ABCA3 TMEM8A LOC107987233 LUC7L KCTD5 CAPN15 LOC105371046 HAGH LOC106660606 MIR3180-5 SNORA64 LOC652276 ECI1 NOXO1 PRSS29P SNHG19 MSRB1 RPL3L CLCN7 RHBDL1 RHOT2 LINC00254 MIR6511B1 TBC1D24 MIR3178 RHBDF1 ZNF598 TSR3 E4F1 SNORA10 LOC101929280 HBA2 C16orf59 MIR1225 MIR6859-4 BAIAP3 FAHD1 FAM173A RPUSD1 SSTR5 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | TBL3 GFER SYNGR3 LOC105371049 BRICD5 NTHL1 TSC2 PKD1 ABCA3 SNORD60 RAB26 RNPS1 LOC106660606 MIR3180-5 SLC9A3R2 ECI1 DNASE1L2 NOXO1 SNHG19 TRAF7 CASKIN1 MIR6511B1 PGP MIR4717 ZNF598 E4F1 NPW MIR3677 MIR940 MIR4516 MIR1225 MLST8 |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss | DNASE1L2 SNHG19 TRAF7 CASKIN1 LOC105371049 MIR6511B1 PGP BRICD5 TSC2 PKD1 E4F1 SNORD60 MIR4516 RAB26 MIR1225 MIR3180-5 MLST8 ECI1 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | DNASE1L2 SNHG19 TRAF7 CASKIN1 PGP BRICD5 E4F1 MIR3677 SNORD60 MIR940 RAB26 RNPS1 LOC106660606 MLST8 ECI1 |
nsv1711 | Chr.16:2199093 - 2232650 on Build GRCh38 | Insertion | PGP BRICD5 E4F1 MLST8 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | MIR940 DNASE1L2 TRAF7 CASKIN1 RNPS1 PGP BRICD5 LOC106660606 E4F1 MIR3677 MLST8 ECI1 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | LOC105371049 BRICD5 LOC729652 TSC2 PKD1 ABCA3 CEMP1 SNORD60 ATP6V0C RAB26 ABCA17P RNPS1 LOC106660606 MIR3180-5 LOC652276 MIR6767 ECI1 DNASE1L2 SNHG19 TRAF7 CASKIN1 MIR6511B1 TBC1D24 PGP MIR4717 MIR3178 E4F1 MIR3677 MIR940 MIR4516 NTN3 C16orf59 PDPK1 MIR1225 CCNF MIR6768 MLST8 AMDHD2 |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | DNASE1L2 SNHG19 TRAF7 CASKIN1 PGP BRICD5 MIR4717 E4F1 ABCA3 MIR3677 SNORD60 MIR940 RAB26 RNPS1 LOC106660606 MLST8 ECI1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |