Assay Details
Target Gene Details
Entrez Gene ID: | 83706 |
Gene Name: | fermitin family member 3 |
Gene Aliases: |
KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF |
Location: |
Chr.11:64206665-64223891 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FERMT3 | NM_031471.5 | 15 | 2304 | NP_113659.3 |
NM_178443.2 | 15 | 2316 | NP_848537.1 | |
XM_011545294.2 | 15 | 2328 | XP_011543596.1 | |
XM_011545295.2 | 14 | 2161 | XP_011543597.1 | |
XM_017018398.1 | 15 | 2312 | XP_016873887.1 | |
XM_017018399.1 | 14 | 2164 | XP_016873888.1 | |
AK093719.1 | 15 | 2249 | BAC04220.1 | |
AY093951.1 | 15 | 2250 | AAM19736.1 | |
AY093952.1 | 15 | 2262 | AAM19737.1 | |
BC004347.1 | 15 | 2248 | AAH04347.2 | |
BC013366.2 | 15 | 2248 | AAH13366.1 | |
BC015584.2 | 15 | 2243 | AAH15584.1 | |
BG491422.1 | 2 | 486 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | MACROD1 PPP1R14B VEGFB TRPT1 PLCB3 LOC105369340 STIP1 NUDT22 GPR137 BAD FERMT3 FKBP2 DNAJC4 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | MACROD1 KCNK4-TEX40 PPP1R14B VEGFB PRDX5 FLRT1 STIP1 NUDT22 FERMT3 DNAJC4 TRMT112 ESRRA COX8A NAA40 CCDC88B TRPT1 PLCB3 LOC105369340 GPR137 BAD KCNK4 FKBP2 TEX40 MIR7155 OTUB1 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | TRMT112 KCNK4-TEX40 ESRRA PPP1R14B VEGFB PRDX5 TRPT1 PLCB3 LOC105369340 STIP1 NUDT22 GPR137 BAD KCNK4 FERMT3 FKBP2 TEX40 DNAJC4 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | LOC105369340 STIP1 NUDT22 PPP1R14B VEGFB FERMT3 FKBP2 DNAJC4 TRPT1 PLCB3 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | TRMT112 KCNK4-TEX40 ESRRA PPP1R14B VEGFB PRDX5 CCDC88B TRPT1 PLCB3 LOC105369340 NUDT22 GPR137 RPS6KA4 BAD KCNK4 FERMT3 FKBP2 TEX40 DNAJC4 MIR1237 MIR7155 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | MACROD1 PPP1R14B VEGFB TRPT1 PLCB3 LOC105369340 STIP1 NUDT22 GPR137 BAD FERMT3 FKBP2 DNAJC4 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | TRMT112 MACROD1 KCNK4-TEX40 ESRRA PPP1R14B VEGFB PRDX5 TRPT1 FLRT1 PLCB3 LOC105369340 STIP1 NUDT22 GPR137 BAD KCNK4 FERMT3 FKBP2 TEX40 DNAJC4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |