Assay Details
Target Gene Details
Entrez Gene ID: | 80194 |
Gene Name: | transmembrane protein 134 |
Gene Aliases: |
- |
Location: |
Chr.11:67464348-67469277 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM134 | NM_001078650.2 | NP_001072118.1 | ||
NM_001078651.2 | NP_001072119.1 | |||
NM_025124.3 | NP_079400.1 | |||
NR_073409.1 | ||||
NR_073410.1 | ||||
NR_073411.1 | ||||
NR_073412.1 | ||||
NR_073413.1 | ||||
XM_006718693.2 | XP_006718756.1 | |||
XM_011545268.2 | XP_011543570.1 | |||
XM_017018356.1 | XP_016873845.1 | |||
AK025402.1 | BAB15126.1 | |||
AK296988.1 | ||||
AK303342.1 | ||||
AK310743.1 | ||||
AY007143.1 | ||||
BC125134.1 | ||||
BG488634.1 | ||||
BG719036.1 | ||||
BX400047.2 | ||||
CN336445.1 | ||||
DN999439.1 | ||||
HY053672.1 | ||||
HY059123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | GPR152 MIR6752 TMEM134 RPS6KB2 PITPNM1 CARNS1 CORO1B PTPRCAP CDK2AP2 CABP4 AIP |
esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication | FAM86C2P MIR6752 C11orf72 TBX10 DOC2GP PITPNM1 CDK2AP2 AIP ACY3 TMEM134 NUDT8 CABP2 GSTP1 ALDH3B2 NDUFV1 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | GPR152 MIR6752 RAD9A PITPNM1 CORO1B PTPRCAP CDK2AP2 CABP4 AIP TMEM134 RPS6KB2 TBC1D10C CABP2 CARNS1 PPP1CA |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | GPR152 MIR6752 RAD9A PITPNM1 CORO1B CLCF1 PTPRCAP CDK2AP2 CABP4 AIP TMEM134 RPS6KB2 TBC1D10C CABP2 CARNS1 PPP1CA LOC100130987 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | GPR152 MIR6752 PITPNM1 CORO1B PTPRCAP CDK2AP2 CABP4 AIP TMEM134 RPS6KB2 TBC1D10C CABP2 CARNS1 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | GPR152 MIR6752 TMEM134 PITPNM1 CABP2 CORO1B CDK2AP2 CABP4 AIP |