Assay Details
Target Gene Details
Entrez Gene ID: | 2782 |
Gene Name: | G protein subunit beta 1 |
Gene Aliases: |
MRD42 |
Location: |
Chr.1:1785285-1891117 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNB1 | NM_001282538.1 | NP_001269467.1 | ||
NM_001282539.1 | NP_001269468.1 | |||
NM_002074.4 | NP_002065.1 | |||
XM_017001059.1 | XP_016856548.1 | |||
XM_017001060.1 | XP_016856549.1 | |||
XM_017001061.1 | XP_016856550.1 | |||
AF070597.1 | AAC28652.1 | |||
AF070603.1 | AAC28655.1 | |||
AF501882.1 | AAM15918.1 | |||
AK096614.1 | ||||
AK122949.1 | ||||
AK312614.1 | ||||
BC004186.2 | AAH04186.1 | |||
BC005888.2 | AAH05888.1 | |||
BC008991.1 | AAH08991.1 | |||
BC114618.1 | AAI14619.1 | |||
BT007305.1 | AAP35969.1 | |||
CR456784.1 | CAG33065.1 | |||
M36430.1 | AAA63265.1 | |||
X04526.1 | CAA28207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5n111 | Chr.1:1575421 - 1823403 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834091 | Chr.1:1701359 - 1880638 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv951380 | Chr.1:1785162 - 1791261 on Build GRCh38 | Deletion |
![]() |
nsv1160797 | Chr.1:1566216 - 1805918 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160926 | Chr.1:1772699 - 1905488 on Build GRCh38 | Duplication |
![]() ![]() ![]() |
dgv10n100 | Chr.1:1779975 - 1884542 on Build GRCh38 | Gain |
![]() ![]() |
esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)