Assay Details
Target Gene Details
Entrez Gene ID: | 9227 |
Gene Name: | lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase) |
Gene Aliases: |
LCA14 |
Location: |
Chr.4:154740841-154753119 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRAT | NM_001301645.1 | 2 | 272 | NP_001288574.1 |
NM_004744.4 | 2 | 466 | NP_004735.2 | |
AF071510.1 | 2 | 305 | AAD13529.1 | |
AK292598.1 | 2 | 466 | ||
AY546085.1 | 1 | 540 | AAS49412.1 | |
AY546086.1 | 2 | 437 | AAS49413.1 | |
BC031053.1 | 2 | 275 | AAH31053.1 | |
DQ266889.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |