Assay Details
Target Gene Details
Entrez Gene ID: | 116369 |
Gene Name: | solute carrier family 26 member 8 |
Gene Aliases: |
SPGF3, TAT1 |
Location: |
Chr.6:35943514-36024855 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC26A8 | NM_001193476.1 | 19 | 2491 | NP_001180405.1 |
NM_052961.3 | 19 | 2515 | NP_443193.1 | |
NM_138718.2 | 17 | 2238 | NP_619732.2 | |
XM_011514294.2 | 18 | 2656 | XP_011512596.1 | |
XM_017010235.1 | 19 | 2541 | XP_016865724.1 | |
AF314959.1 | 18 | 2385 | AAL26868.1 | |
AF331522.1 | 19 | 2459 | AAK95666.1 | |
AK055314.1 | 9 | 1961 | ||
AK057276.1 | 9 | 1366 | BAB71408.1 | |
AK097410.1 | 13 | 1577 | ||
AK122928.1 | 19 | 2491 | ||
BC025408.2 | 17 | 2231 | AAH25408.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |